Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4-tert-butylphenol 98%

Supplier: AMBEED, INC
Warning

A502531-100G A502531-25G A502531-5G A502531-500G
103839-394EA 6 USD InStock
103839-394 76695-716 76843-362 76843-364 77880-089
2-Amino-4-tert-butylphenol 98%
2-Amino-4-tert-butylphenol
Precaución: for Research Use Only


Formula: C₁₀H₁₅NO
MW: 165.24 g/mol
Melting Pt: 160…163 °C
Flash Pt: 120.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00007698
CAS Number: 1199-46-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty