Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxy-4-(trifluoromethyl)aniline 95%

Supplier: AMBEED, INC
Warning

A134170-1G A134170-10G A134170-250MG A134170-5G
103801-980EA 47 USD InStock
103801-980 77258-562 77258-564 77314-698
2-Hydroxy-4-(trifluoromethyl)aniline 95%
2-Hydroxy-4-(trifluoromethyl)aniline
Precaución: for Research Use Only


Formula: C₇H₆F₃NO
Boiling Pt: 233.8 °C (760 mm Hg)
Flash Pt: 95.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08056279
CAS Number: 454-82-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty