Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-fluorophenol 98%

Supplier: AMBEED, INC
Warning

A149105-5G A149105-100G A149105-10G A149105-25G A149105-5G
103842-122EA 14 USD InStock
103842-122 77286-058 77286-060 77317-600 77317-602
2-Amino-5-fluorophenol 98%
2-Amino-5-fluorophenol
Precaución: for Research Use Only


Formula: C₆H₆FNO
MW: 127.12
Boiling Pt: 246.1 °C (760 mm Hg)
Melting Pt: 38 °C
Flash Pt: 102.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00671759
CAS Number: 53981-24-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty