Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-hydroxybenzoic acid 95%

Supplier: AMBEED, INC
Warning

A127036-100G A127036-10G A127036-500G A127036-25G A127036-5G
77189-920EA 65 USD InStock
77189-920 77189-922 77189-924 77313-150 77313-152
2-Amino-5-hydroxybenzoic acid 95%
2-Amino-5-hydroxybenzoic acid
Precaución: for Research Use Only


Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 347
Melting Pt: 247 °C
Flash Pt: 206.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00007870
CAS Number: 394-31-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty