Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-6-bromobenzoxazole 98%

Supplier: AMBEED, INC
Warning

A147406-1G A147406-5G A147406-100MG A147406-250MG A147406-1G
103803-770EA 22 USD InStock
103803-770 103841-872 76812-038 76812-040 77317-244
2-Amino-6-bromobenzoxazole 98%
2-Amino-6-bromobenzoxazole


Formula: C₇H₅BrN₂O
Boiling Pt: 340.4 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD01664214
CAS Number: 52112-66-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty