Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-6-bromonicotinic acid 97%

Supplier: AMBEED, INC
Warning

A133712-1G A133712-100MG A133712-250MG A133712-1G A133712-5G
103801-902EA 70 USD InStock
103801-902 76811-360 76811-362 77298-650 77298-652
2-Amino-6-bromonicotinic acid 97%
2-Amino-6-bromonicotinic acid
Precaución: for Research Use Only


Formula: C₆H₅BrN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD13190078
CAS Number: 1196157-51-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty