Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-6-bromopyridin-3-ol 95%

Supplier: AMBEED, INC
Warning

A174045-1G A174045-5G A174045-250MG A174045-5G A174045-1G
103806-908EA 42 USD InStock
103806-908 103845-206 76813-314 77082-490 77301-312
2-Amino-6-bromopyridin-3-ol 95%
2-Amino-6-bromopyridin-3-ol
Precaución: for Research Use Only


Formula: C₅H₅BrN₂O
MW: 189.01 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11110242
CAS Number: 934758-27-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty