Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-6-chlorobenzonitrile 98%

Supplier: AMBEED, INC
Warning

A232229-1G A232229-5G A232229-25G A232229-5G A232229-100G A232229-10G A232229-1G
103810-824EA 9 USD InStock
103810-824 103850-576 77089-504 77089-506 77261-570 77261-572 77303-934
2-Amino-6-chlorobenzonitrile 98%
2-Amino-6-chlorobenzonitrile
Precaución: for Research Use Only


Formula: C₇H₅ClN₂
MW: 152.58
Boiling Pt: 301
Melting Pt: 134…136 °C
Flash Pt: 151.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00015546
CAS Number: 6575-11-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty