Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-6-chloropurine 98%

Supplier: AMBEED, INC
Warning

A135577-1000G A135577-100G A135577-10G A135577-500G A135577-5G A135577-25G
103808-510EA 177 USD InStock
103808-510 77272-218 77272-220 77272-222 77272-224 77298-798
2-Amino-6-chloropurine 98%
2-Amino-6-chloropurine
Precaución: for Research Use Only


Formula: C₅H₄ClN₅
MW: 169.57 g/mol
Boiling Pt: 623.9 °C (760 mm Hg)
Melting Pt: 300
Flash Pt: 144.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00075252
CAS Number: 10310-21-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty