Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Nitroanthranilic acid 97%

Supplier: AMBEED, INC
Warning

A356003-1G A356003-25G A356003-5G A356003-100G A356003-10G
103826-158EA 6 USD InStock
103826-158 103837-166 76840-296 76840-298 76840-300 77263-922 77746-699
6-Nitroanthranilic acid 97%
6-Nitroanthranilic acid
Precaución: for Research Use Only


Formula: C₇H₆N₂O₄
MW: 182.14 g/mol
Boiling Pt: 412.3 °C (760 mm Hg)
Melting Pt: 179…180 °C
Flash Pt: 203.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00034785
CAS Number: 50573-74-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty