Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Aminobenzo[d]thiazol-4-ol 97%

Supplier: AMBEED, INC
Warning

A102595-1G A102595-5G A102595-250MG A102595-25G
103796-394EA 30 USD InStock
103796-394 103822-090 77245-194 77245-196 77245-198
2-Aminobenzo[d]thiazol-4-ol 97%
2-Aminobenzo[d]thiazol-4-ol
Precaución: for Research Use Only


Formula: C₇H₆N₂OS
MW: 166.2003 g/mol
Boiling Pt: 394.6 °C (760 mm Hg)
Melting Pt: 179…181 °C
Storage Temperature: Ambient
MDL Number: MFCD00160060
CAS Number: 7471-03-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty