Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Aminobenzo[d]thiazole-6-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A125455-1G A125455-100G A125455-10G A125455-250MG A125455-25G A125455-1G A125455-5G
103800-680EA 15 USD InStock
103800-680 77258-266 77258-268 77258-270 77258-272 77312-826 77312-828
2-Aminobenzo[d]thiazole-6-carboxylic acid 97%
2-Aminobenzo[d]thiazole-6-carboxylic acid
Precaución: for Research Use Only


Formula: C₈H₆N₂O₂S
Boiling Pt: 466.6 °C (760 mm Hg)
Melting Pt: 265 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00054180
CAS Number: 93-85-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty