Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Aminocyclohexanol 97%

Supplier: AMBEED, INC
Danger

A117930-1G A117930-10G A117930-25G A117930-1G A117930-5G
103798-956EA 9 USD InStock
103798-956 103824-846 77148-996 77148-998 77149-000 77311-626 77311-628
2-Aminocyclohexanol 97%
2-Aminocyclohexanol
Precaución: For research use only.


Formula: C₆H₁₃NO
Boiling Pt: 212 °C (760 mm Hg)
Melting Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00191368
CAS Number: 6850-38-0
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty