Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Aminocyclopentanol 98%

Supplier: AMBEED, INC
Danger

A198485-5G A198485-1G A198485-1MG A198485-10G
103817-024EA 43 USD InStock
103817-024 103847-650 77085-680 77112-078 77685-495
2-Aminocyclopentanol 98%
2-Aminocyclopentanol
Precaución: for Research Use Only


Formula: C₅H₁₁NO
Flash Pt: 62.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00100903
CAS Number: 89381-13-5
UN: 3082
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty