Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-3-furonitrile 95%

Supplier: AMBEED, INC
Warning

A477407-1G A477407-250MG A477407-5G
103828-096EA 537 USD InStock
103828-096 103839-168 76818-594 76843-102
2-Amino-3-furonitrile 95%
2-Amino-3-furonitrile
Precaución: for Research Use Only


Formula: C₅H₄N₂O
Storage Temperature: Refrigerator
MDL Number: MFCD09834958
CAS Number: 139370-56-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty