Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Aminoheptanoic acid 97%

Supplier: AMBEED, INC
Warning

A174495-1G A174495-5G A174495-100MG A174495-250MG A174495-1G A174495-5G
103806-948EA 55 USD InStock
103806-948 103845-248 77111-130 77111-132 77301-336 77301-338
2-Aminoheptanoic acid 97%
2-Aminoheptanoic acid
Precaución: for Research Use Only


Formula: C₇H₁₅NO₂
MW: 145.2 g/mol
Melting Pt: 280 °C
Storage Temperature: Ambient
MDL Number: MFCD00136760
CAS Number: 1115-90-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty