Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Aminophthalimide 97%

Supplier: AMBEED, INC
Warning

A113280-1G A113280-100G A113280-10G A113280-25G A113280-500G A113280-1G
103798-206EA 5 USD InStock
103798-206 103824-046 77233-708 77233-710 77233-712 77233-714 77297-236
N-Aminophthalimide 97%
N-Aminophthalimide
Precaución: for Research Use Only


Formula: C₈H₆N₂O₂
Boiling Pt: 348.6 °C (760 mm Hg)
Melting Pt: 200…202 °C
Flash Pt: 164.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00005895
CAS Number: 1875-48-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty