Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-N-(2,2,2-trifluoroethyl)acetamide hydrochloride 95%

Supplier: AMBEED, INC
Warning

A104389-1G A104389-5G A104389-10G A104389-250MG A104389-25G A104389-1G A104389-5G
103796-702EA 12 USD InStock
103796-702 103822-416 77160-378 77160-380 77160-382 77309-282 77309-284
2-Amino-N-(2,2,2-trifluoroethyl)acetamide hydrochloride 95%
2-Amino-N-(2,2,2-trifluoroethyl)acetamide hydrochloride
Precaución: for Research Use Only


Formula: C₄H₇N₂OF₃·HCl
Storage Temperature: Ambient
MDL Number: MFCD09944516
CAS Number: 1171331-39-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty