Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Aminopent-4-ynoic acid hydrochloride 97%

Supplier: AMBEED, INC
Warning

A420478-1G A420478-5G A420478-250MG A420478-1G
103827-386EA 77 USD InStock
103827-386 103838-434 77143-146 77307-206
2-Aminopent-4-ynoic acid hydrochloride 97%
2-Aminopent-4-ynoic acid hydrochloride
Precaución: for Research Use Only


Formula: C₅H₈ClNO₂
MW: 149.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11113137
CAS Number: 16900-57-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty