Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-2-Amino-1-propanol 98%

Supplier: AMBEED, INC
Warning

A131092-100G A131092-10G A131092-500G A131092-25G
77258-468EA 69 USD InStock
77258-468 77258-470 77258-472 77298-448
(±)-2-Amino-1-propanol 98%
(±)-2-Amino-1-propanol
Precaución: for Research Use Only


Formula: C₃H₉NO
MW: 75.11 g/mol
Boiling Pt: 173
Melting Pt: 8
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00008085
CAS Number: 6168-72-5
Merck Index: 12,00488
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty