Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Aminothiazole-5-carbaldehyde hydrochloride 95%

Supplier: AMBEED, INC
Warning

A288194-1G A288194-5G A288194-100MG A288194-250MG
103813-784EA 23 USD InStock
103813-784 103834-926 77250-612 77250-614
2-Aminothiazole-5-carbaldehyde hydrochloride 95%
2-Aminothiazole-5-carbaldehyde hydrochloride
Precaución: for Research Use Only


Formula: C₄H₅ClN₂OS
MW: 164.6100 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09991571
CAS Number: 920313-27-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty