Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Benzofuranacetic acid 96%

Supplier: AMBEED, INC
Warning

A280999-1G A280999-5G A280999-250MG A280999-100MG
103813-530EA 87 USD InStock
103813-530 103853-482 77262-652 77746-505
2-Benzofuranacetic acid 96%
2-Benzofuranacetic acid


Formula: C₁₀H₈O₃
Boiling Pt: 331 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00052742
CAS Number: 62119-70-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty