Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Benzyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%

Supplier: AMBEED, INC
Warning

A247225-1G A247225-5G A247225-10G A247225-1G A247225-5G A247225-25G
103811-758EA 5 USD InStock
103811-758 103851-592 76692-402 77090-916 77090-918 77304-402
2-Benzyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
2-Benzyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only


Formula: C₁₃H₁₉BO₂
Boiling Pt: 269.6 °C (760 mm Hg)
Flash Pt: 116.9 °C
Storage Temperature: Freezer
MDL Number: MFCD05663841
CAS Number: 87100-28-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty