Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

ω-Bromo-4-fluoroacetophenone 98%

Supplier: AMBEED, INC
Danger

A902511-5G A902511-25G A902511-10G A902511-5G
103841-352EA 5 USD InStock
103841-352 76845-622 77244-730 77308-498
ω-Bromo-4-fluoroacetophenone 98%
ω-Bromo-4-fluoroacetophenone
Precaución: for Research Use Only


Formula: C₈H₆BrFO
MW: 217.04 g/mol
Boiling Pt: 82
Melting Pt: 49
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00040830
CAS Number: 403-29-2
UN: 3261
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty