Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

ω-Bromo-4-iodoacetophenone 95%

Supplier: AMBEED, INC
Warning

A113911-1G A113911-1G A113911-5G A113911-10G A113911-25G
103798-318EA 6 USD InStock
103798-318 103824-160 77310-976 77310-978 77570-758 77685-009
ω-Bromo-4-iodoacetophenone 95%
ω-Bromo-4-iodoacetophenone
Precaución: for Research Use Only


Formula: C₈H₆BrIO
Boiling Pt: 328.7 °C (760 mm Hg)
Flash Pt: 152.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01360521
CAS Number: 31827-94-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty