Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-10H-phenothiazine 95%

Supplier: AMBEED, INC
Warning

A164195-50MG A164195-1G A164195-100MG A164195-5G A164195-1G A164195-50MG
103799-404EA 39 USD InStock
103799-404 103805-824 77259-730 77259-732 77300-696 77300-698
2-Bromo-10H-phenothiazine 95%
2-Bromo-10H-phenothiazine
Precaución: for Research Use Only


Formula: C₁₂H₈BrNS
Boiling Pt: 408.4 °C (760 mm Hg)
Flash Pt: 200.8 °C
Storage Temperature: Ambient
MDL Number: MFCD27921901
CAS Number: 66820-95-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty