Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-3-fluorotoluene 98%

Supplier: AMBEED, INC
Warning

A120454-1G A120454-10G A120454-25G A120454-500G A120454-1G A120454-5G
103799-936EA 9 USD InStock
103799-936 77136-476 77136-478 77136-480 77312-040 77312-042
2-Bromo-3-fluorotoluene 98%
2-Bromo-3-fluorotoluene
Precaución: for Research Use Only


Formula: C₇H₆BrF
Boiling Pt: 182.8 °C (760 mm Hg)
Flash Pt: 66.7 °C
Storage Temperature: Ambient
MDL Number: MFCD08458010
CAS Number: 59907-13-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty