Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-3-(trifluoromethoxy)pyridine 95%

Supplier: AMBEED, INC
Warning

A689099-5G A689099-100MG A689099-250MG A689099-5G
103840-570EA 1440 USD InStock
103840-570 76819-984 76819-986 77308-122
2-Bromo-3-(trifluoromethoxy)pyridine 95%
2-Bromo-3-(trifluoromethoxy)pyridine
Precaución: for Research Use Only


Formula: C₆H₃BrF₃NO
MW: 242 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20486158
CAS Number: 1206978-11-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty