Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-3,5-dichloropyridine 98%

Supplier: AMBEED, INC
Warning

A392632-100G A392632-10G A392632-25G
103837-940EA 11 USD InStock
103837-940 77278-650 77278-652 77307-010
2-Bromo-3,5-dichloropyridine 98%
2-Bromo-3,5-dichloropyridine
Precaución: for Research Use Only


Formula: C₅H₂BrCl₂N
Boiling Pt: 243.3 °C (760 mm Hg)
Melting Pt: 40…42 °C
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00233990
CAS Number: 14482-51-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty