Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-3-iodobenzoic acid 96%

Supplier: AMBEED, INC
Warning

A181786-1G A181786-5G A181786-250MG A181786-1G
103807-648EA 33 USD InStock
103807-648 103846-022 76824-944 77083-546
2-Bromo-3-iodobenzoic acid 96%
2-Bromo-3-iodobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₄BrIO₂
MW: 326.914 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11036150
CAS Number: 855198-37-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty