Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-(pyrrolidin-1-yl)pyridine 97%

Supplier: AMBEED, INC
Warning

A124802-1G A124802-250MG A124802-1G
103800-580EA 98 USD InStock
103800-580 77234-300 77312-722
2-Bromo-4-(pyrrolidin-1-yl)pyridine 97%
2-Bromo-4-(pyrrolidin-1-yl)pyridine
Precaución: for Research Use Only


Formula: C₉H₁₁BrN₂
Boiling Pt: 335.5±27.0 °C (760 mm Hg)
Melting Pt: 124 °C
Flash Pt: 156.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09032268
CAS Number: 230618-42-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty