Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-(tert-butyl)benzoic acid 98%

Supplier: AMBEED, INC
Warning

A216267-1G A216267-5G A216267-250MG A216267-1G A216267-5G
103818-550EA 68 USD InStock
103818-550 103849-266 76826-048 77303-432 77303-434
2-Bromo-4-(tert-butyl)benzoic acid 98%
2-Bromo-4-(tert-butyl)benzoic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₃BrO₂
MW: 257.12 g/mol
Boiling Pt: 310.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD11040238
CAS Number: 6332-96-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty