Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-(trifluoromethyl)aniline 98%

Supplier: AMBEED, INC
Warning

A103261-5G A103261-100G A103261-10G A103261-25G A103261-500G A103261-5G
103822-224EA 6 USD InStock
103822-224 76765-294 76765-296 76765-298 76765-300 77283-072
2-Bromo-4-(trifluoromethyl)aniline 98%
2-Bromo-4-(trifluoromethyl)aniline
Precaución: for Research Use Only


Formula: C₇H₅BrF₃N
MW: 240.02
Boiling Pt: 219.7 °C (760 mm Hg)
Melting Pt: 26…28 °C
Density: 1.7
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00042150
CAS Number: 57946-63-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty