Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4,5-dichloropyridine 95%

Supplier: AMBEED, INC
Warning

A304181-1G A304181-1G A304181-5G A304181-100MG A304181-250MG
103814-412EA 225 USD InStock
103814-412 103835-576 76838-268 76838-270 76875-682 76875-684
2-Bromo-4,5-dichloropyridine 95%
2-Bromo-4,5-dichloropyridine
Precaución: for Research Use Only


Formula: C₅H₂BrCl₂N
Boiling Pt: 240.6±35.0 °C (760 mm Hg)
Flash Pt: 99.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11100656
CAS Number: 1033203-42-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty