Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4,6-difluoroanisole 98%

Supplier: AMBEED, INC
Warning

A124204-1G A124204-10G A124204-1G A124204-5G A124204-100G
103800-504EA 5 USD InStock
103800-504 76766-778 77312-636 77312-638 77876-898
2-Bromo-4,6-difluoroanisole 98%
2-Bromo-4,6-difluoroanisole
Precaución: for Research Use Only


Formula: C₇H₅BrF₂O
Boiling Pt: 212.9 °C (760 mm Hg)
Flash Pt: 97.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00070754
CAS Number: 202865-59-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty