Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-chloro-3-nitropyridine 95%

Supplier: AMBEED, INC
Warning

A139416-1G A139416-100MG A139416-250MG A139416-1G
103802-736EA 183 USD InStock
103802-736 76647-002 76647-004 77315-830
2-Bromo-4-chloro-3-nitropyridine 95%
2-Bromo-4-chloro-3-nitropyridine
Precaución: for Research Use Only


Formula: C₅H₂BrClN₂O₂
MW: 237.44 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17014974
CAS Number: 1261732-45-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty