Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-chloro-5-nitropyridine 95%

Supplier: AMBEED, INC
Warning

A125784-1G A125784-100MG A125784-250MG
103800-724EA 248 USD InStock
103800-724 76983-462 76983-464
2-Bromo-4-chloro-5-nitropyridine 95%
2-Bromo-4-chloro-5-nitropyridine
Precaución: for Research Use Only


Formula: C₅H₂BrClN₂O₂
MW: 237.44 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17014975
CAS Number: 1137475-57-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty