Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-fluorophenol 99%

Supplier: AMBEED, INC
Warning

A135776-100G A135776-10G A135776-500G A135776-5G
77285-496EA 47 USD InStock
77285-496 77285-498 77285-500 77298-818
2-Bromo-4-fluorophenol 99%
2-Bromo-4-fluorophenol
Precaución: for Research Use Only


Formula: C₆H₄BrFO
Boiling Pt: 260.6 °C (760 mm Hg)
Melting Pt: 43…45 °C
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00010614
CAS Number: 496-69-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty