Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-methylpentanoic acid 97%

Supplier: AMBEED, INC
Danger

A465544-5G A465544-1G A465544-5G A465544-100MG A465544-250MG
103827-972EA 26 USD InStock
103827-972 103839-046 76842-954 76842-956 76979-594 76979-596
2-Bromo-4-methylpentanoic acid 97%
2-Bromo-4-methylpentanoic acid
Precaución: For research use only


Formula: C₆H₁₁BrO₂
Boiling Pt: 241 °C (760 mm Hg)
Flash Pt: 99 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039606
CAS Number: 42990-24-9
UN: 3265
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty