Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-methylthiazole-5-carbaldehyde 95%

Supplier: AMBEED, INC
Warning

A124003-1G A124003-100MG A124003-250MG A124003-1G A124003-5G
103800-470EA 143 USD InStock
103800-470 77136-624 77136-626 77136-628 77312-600 77312-602
2-Bromo-4-methylthiazole-5-carbaldehyde 95%
2-Bromo-4-methylthiazole-5-carbaldehyde
Precaución: for Research Use Only


Formula: C₅H₄BrNOS
MW: 206.06 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22041605
CAS Number: 933720-87-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty