Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-nitroaniline 97%

Supplier: AMBEED, INC
Warning

A474356-1G A474356-5G A474356-25G A474356-5G A474356-100G A474356-10G A474356-500G A474356-1G
103828-060EA 9 USD InStock
103828-060 103839-134 76843-062 76843-064 77156-366 77156-368 77156-370 77307-512
2-Bromo-4-nitroaniline 97%
2-Bromo-4-nitroaniline
Precaución: for Research Use Only


Formula: C₆H₅BrN₂O₂
MW: 217.02
Boiling Pt: 351.8 °C (760 mm Hg)
Melting Pt: 104 °C
Flash Pt: 166.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00025152
CAS Number: 13296-94-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty