Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-phenylpyridine 97%

Supplier: AMBEED, INC
Warning

A185231-1G A185231-5G A185231-1G A185231-25G A185231-5G
103815-834EA 16 USD InStock
103815-834 103846-402 77260-356 77301-958 77301-960 77301-962
2-Bromo-4-phenylpyridine 97%
2-Bromo-4-phenylpyridine
Precaución: for Research Use Only


Formula: C₁₁H₈BrN
Storage Temperature: Ambient
MDL Number: MFCD00235159
CAS Number: 54151-74-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty