Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-tert-butylphenol 98%

Supplier: AMBEED, INC
Warning

A261787-1G A261787-5G A261787-25G A261787-100G A261787-10G A261787-500G A261787-1G A261787-5G
103812-534EA 5 USD InStock
103812-534 103852-424 77092-032 77262-246 77262-248 77262-250 77304-824 77304-826
2-Bromo-4-tert-butylphenol 98%
2-Bromo-4-tert-butylphenol
Precaución: for Research Use Only


Formula: C₁₀H₁₃BrO
Boiling Pt: 245.9 °C (760 mm Hg)
Flash Pt: 102.5 °C
Storage Temperature: Ambient
MDL Number: MFCD02682891
CAS Number: 2198-66-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty