Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-5-(difluoromethoxy)pyridine 95%

Supplier: AMBEED, INC
Warning

A100832-1G A100832-5G A100832-100MG A100832-250MG A100832-1G A100832-5G A100832-10G
103795-648EA 42 USD InStock
103795-648 103821-344 76869-118 76869-120 77296-854 77296-856 77670-460
2-Bromo-5-(difluoromethoxy)pyridine 95%
2-Bromo-5-(difluoromethoxy)pyridine
Precaución: for Research Use Only


Formula: C₆H₄BrF₂NO
MW: 224.003 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13185842
CAS Number: 845827-14-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty