Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-5-(trifluoromethyl)thiazole 95%

Supplier: AMBEED, INC
Warning

A423755-1G A423755-100MG A423755-250MG A423755-1G A423755-5G
103827-440EA 151 USD InStock
103827-440 103838-490 76818-056 76818-058 76842-128 76842-130
2-Bromo-5-(trifluoromethyl)thiazole 95%
2-Bromo-5-(trifluoromethyl)thiazole
Precaución: for Research Use Only


Formula: C₄HBrF₃NS
MW: 232.022 g/mol
Boiling Pt: 182.8±35.0 °C (760 mm Hg)
Flash Pt: 64.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD14702723
CAS Number: 1209458-80-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty