Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-5-chloro-3-fluoropyridine 98%

Supplier: AMBEED, INC
Warning

A201080-1G A201080-5G A201080-100G A201080-100MG A201080-10G A201080-250MG A201080-1G A201080-25G A201080-5G
103817-264EA 8 USD InStock
103817-264 103847-900 77030-128 77030-130 77030-132 77030-134 77085-974 77085-976 77085-978
2-Bromo-5-chloro-3-fluoropyridine 98%
2-Bromo-5-chloro-3-fluoropyridine
Precaución: for Research Use Only


Formula: C₅H₂BrClFN
Boiling Pt: 190.8 °C (760 mm Hg)
Flash Pt: 69.185 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11044268
CAS Number: 514797-97-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty