Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-5-iodo-1,3-dimethylbenzene 97%

Supplier: AMBEED, INC
Warning

A422125-1G A422125-25G A422125-5G A422125-100G A422125-10G A422125-250MG
103827-414EA 13 USD InStock
103827-414 103838-462 76842-084 76842-086 76842-088 76990-448 76990-450 76990-452
2-Bromo-5-iodo-1,3-dimethylbenzene 97%
2-Bromo-5-iodo-1,3-dimethylbenzene
Precaución: for Research Use Only


Formula: C₈H₈BrI
Boiling Pt: 282.4 °C (760 mm Hg)
Flash Pt: 125 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07780650
CAS Number: 689260-53-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty