Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-5-methylbenzaldehyde 98%

Supplier: AMBEED, INC
Warning

A293419-1G A293419-1G A293419-25G A293419-5G A293419-100G A293419-10G A293419-100MG
103813-982EA 9 USD InStock
103813-982 103835-120 76837-738 76837-740 76837-742 77032-354 77032-356 77872-377
2-Bromo-5-methylbenzaldehyde 98%
2-Bromo-5-methylbenzaldehyde
Precaución: for Research Use Only


Formula: C₈H₇BrO
Melting Pt: 41…45 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08669550
CAS Number: 90221-55-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty