Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-5-nitroaniline 98%

Supplier: AMBEED, INC
Warning

A130232-1G A130232-100G A130232-10G A130232-1G A130232-25G A130232-5G
103801-402EA 5 USD InStock
103801-402 77094-938 77094-940 77313-832 77313-834 77313-836
2-Bromo-5-nitroaniline 98%
2-Bromo-5-nitroaniline
Precaución: for Research Use Only


Formula: C₆H₅BrN₂O₂
Melting Pt: 139…141 °C
Flash Pt: 156.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00051578
CAS Number: 10403-47-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty