Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-5-nitropyrimidine 97%

Supplier: AMBEED, INC
Warning

A517869-5G A517869-10G A517869-250MG A517869-1G A517869-5G
103828-482EA 102 USD InStock
103828-482 103839-586 76790-070 76790-072 76843-590 76843-592
2-Bromo-5-nitropyrimidine 97%
2-Bromo-5-nitropyrimidine
Precaución: for Research Use Only


Formula: C₄H₂BrN₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD07778612
CAS Number: 1210824-80-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty